Digoxigenin (DIG) alkyne, FastClick™
Lieferant: AAT BIOQUEST
72900
AATB72900EA 266 EUR
AATB72900
Digoxigenin (DIG) alkyne, FastClick™
Digoxigenin (DIG) alkyne
FastClick™ Digoxigenin (DIG) Alkyne contains both the CAG moiety of FastClick (for assisting click efficiency) and DIG hapten (as the detection tag) for developing DIG-based probes.
  • Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only