Biotin Alkyne, FastClick™
Lieferant: AAT BIOQUEST
FastClick™ Biotin Alkyne contains both the CAG moiety of FastClick (for assisting click efficiency) and biotin (as the detection tag) for developing biotin-based probes.
- Stable for 24 months
Quality tested in-house
Formel:
C₁₃H₁₉N₃O₂S |
MDL:
MFCD28385487 CAS-Nummer: 773888-45-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...