Sulphorhodamine G ≥95%

Lieferant: AAT BIOQUEST
Achtung

73
AATB73EA 120 EUR
AATB73
Sulphorhodamine G ≥95%
Sulphorhodamine G

Sulforhodamine G is used as a polar tracer as sulforhodamine B.

MDL: MFCD00213704
CAS-Nummer: 5873-16-5
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR