Rhodamine 101 inner salt ≥95%
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
9-(2-Carboxyphenyl)-2,3,6,7,12,13,16,17-octahydro-1H,5H,11H,15H-Xantheno[2,3,4-ij:5,6,7-i'j′]diquinolizin-18-ium inner salt
74
AATB74EA
120
EUR
AATB74
Rhodamine 101 inner salt ≥95%
Rhodamine 101 inner salt
Rhodamine dyes are used extensively in a variety of biological applications such as fluorescence microscopy, flow cytometry, fluorescence correlation spectroscopy and ELISA.
Formel:
C₃₂H₃₀N₂O₃ Molecular Weight: 490,603 g/mol |
MDL:
MFCD07809431 CAS-Nummer: 116450-56-7 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...