Rhodamine 101 Chloride Salt ≥95%

Lieferant: AAT BIOQUEST
Achtung

75
AATB75EA 120 EUR
AATB75
Rhodamine 101 Chloride Salt ≥95%
Rhodamine 101 Chloride Salt

Rhodamine dyes are used extensively in a variety of biological applications such as fluorescence microscopy, flow cytometry, fluorescence correlation spectroscopy and ELISA.

Formel: C₃₂H₃₁ClN₂O₃
Molecular Weight: 527,05 g/mol
CAS-Nummer: 64339-18-0
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR