Sulphorhodamine 101 ≥95%

Lieferant: AAT BIOQUEST

Synonyme: Sulforhodamine 101, Texas Red, SR 101, Sulforhodamine 640, 9-(2,4-Disulphophenyl)-2,3,6,7,12,13,16,17-octahydro-1H,5H,11H,15H-xantheno[2,3,4-ij:5,6,7-i'j']diquinolizin-18-ium inner salt

77
AATB77EA 114 EUR
AATB77
Sulphorhodamine 101 ≥95%
Sulphorhodamine 101

Sulforhodamine 101 is a fluorescence reference standard for measuring fluorescence quantum yield.


  • Fluorescence Reference Standard


Bitte beachten For research use only.

Formel: C₃₁H₃₀N₂O₇S₂
Molecular Weight: 606,708 g/mol
MDL: MFCD00012407
CAS-Nummer: 60311-02-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR