(-)-Blebbistatin ≥95%
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
(S)-3a-Hydroxy-6-methyl-1-phenyl-1,2,3,3a-tetrahydro-4H-pyrrolo[2,3-b]quinolin-4-one
80001
AATB80001EA
152
EUR
AATB80001
(-)-Blebbistatin ≥95%
(-)-Blebbistatin
(-)-Blebbistatin is a cell-permeable inhibitor for non muscle myosin II ATPase with IC50 of 2 µM.
Formel:
C₁₈H₁₆N₂O₂ Molecular Weight: 292,3318 g/mol |
MDL:
MFCD08460907 CAS-Nummer: 856925-71-8 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...