Staurosporine 1 mM, ReadiUse™ Kontroll-DNS
Lieferant: AAT Bioquest
|
Achtung
|
80050
AATB80050EA
152
EUR
AATB80050
Staurosporine 1 mM, ReadiUse™ Kontroll-DNS
Staurosporine
Staurosporine is widely used as a positive control for inducing apoptosis.
- Well-cited positive apoptosis control
- Simple effective apoptosis induction in multiple cell lines
- Cell permeable and reversible
- Inhibits some types of platelet aggregation
Formel:
C₂₈H₂₆N₄O₃ |
MDL:
MFCD18252446 CAS-Nummer: 62996-74-1 ADR: 5,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...