5-Dimethylaminonaphthalene-1-(N-(5-aminopentyl))Sulphonamide (Dansyl cadaverine) ≥95%

Lieferant: AAT BIOQUEST

Synonyme: Dansylcadaverine, N-(5-Aminopentyl)-5-(dimethylamino)naphthalene-1-sulphonamide

810
AATB810EA 120 EUR
AATB810
5-Dimethylaminonaphthalene-1-(N-(5-aminopentyl))Sulphonamide (Dansyl cadaverine) ≥95%
5-Dimethylaminonaphthalene-1-(N-(5-aminopentyl))Sulphonamide (Dansyl cadaverine)

Dansyl cadaverine is an amine-containing Dansyl building block.


  • Multiple Bioconjugation Applications
  • 95% Purity


Bitte beachten For research use only.

Formel: C₁₇H₂₅N₃O₂S
Molecular Weight: 335,46 g/mol
MDL: MFCD00042704
CAS-Nummer: 10121-91-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR