Dansylchlorid ≥95%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: 5-(Dimethylamino)naphthalinsulfonylchlorid, 5-(Dimethylamino)naphthalinsulfonsäurechlorid, 5-(Dimethylamino)naphthalin-1-sulfonylchlorid

811
AATB811EA 120 EUR
AATB811
Dansylchlorid ≥95%
Dansylchlorid

Dansyl chloride is non-fluorescent until it reacts with amines.

Formel: C₁₂H₁₂ClNO₂S
Molecular Weight: 269,75 g/mol
MDL: MFCD00003985
CAS-Nummer: 605-65-2
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR