7-Hydroxy-4-(trifluormethyl)-2H-chromen-2-on

Lieferant: AAT BIOQUEST
Achtung

Synonyme: 4-(Trifluoromethyl)umbelliferone, TFMU

82
AATB82EA 120 EUR
AATB82
7-Hydroxy-4-(trifluormethyl)-2H-chromen-2-on
7-Hydroxy-4-(trifluoromethyl)coumarin

This coumarin compound is used as a calibration standard for 7-hydroxy-4-trifluoromethylcoumarin-based enzyme substrates.

Formel: C₁₀H₅F₃O₃
Molecular Weight: 230,14 g/mol
MDL: MFCD00037578
CAS-Nummer: 575-03-1
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR