Fluorescamin

Lieferant: AAT BIOQUEST
Achtung

Synonyme: Fluram, 4-Phenyl-3H,3'H-spiro[furan-2,1'-isobenzofuran]-3,3'-dione, 4-Phenylspiro[furan-2(3H),1'-phthalan]-3,3'-dione

820
AATB820EA 152 EUR
AATB820
Fluorescamin
Fluorescamine

Fluorescamine is a popular amine-fluorogenic reagent for determining protein concentrations in solutions and on gels.

Formel: C₁₇H₁₀O₄
Molecular Weight: 278,26 g/mol
MDL: MFCD00005928
CAS-Nummer: 38183-12-9
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR