4-Fluor-7-nitrobenzofurazan
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
4-Fluoro-7-nitro-[2,1,3]-benzoxadiazole, NBD-F
821
AATB821EA
152
EUR
AATB821
4-Fluor-7-nitrobenzofurazan
4-Fluor-7-nitrobenzofurazan
NBD-F has properties and applications similar to those of NBD-Cl.
Formel:
C₆H₂FN₃O₃ Molecular Weight: 183,1 g/mol |
MDL:
MFCD00010196 CAS-Nummer: 29270-56-2 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...