4-Chlor-7-nitrobenzofurazan

Lieferant: AAT BIOQUEST
Achtung

Synonyme: NBD chloride, 4-Chloro-7-Nitrobenz-2-Oxa-1,3-Diazole, NBD-Cl

825
AATB825EA 120 EUR
AATB825
4-Chlor-7-nitrobenzofurazan
4-Chlor-7-nitrobenzofurazan

NBD-Cl is widely used to label peptides, proteins, drugs and other biomolecules.

Formel: C₆H₂ClN₃O₃
Molecular Weight: 199,55 g/mol
MDL: MFCD00005808
CAS-Nummer: 10199-89-0
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR