NBD-X ≥95%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: 6-(N-(7-Nitrobenz-2-oxa-1,3-diazol-4-yl)amino)hexanoic acid, 6-(7-Nitrobenzofurazan-4-ylamino)-hexanoic acid

828
AATB828EA 1170 EUR
AATB828
NBD-X ≥95%
NBD-X

Compared to NBD chloride and fluoride, NBD-X acid gives better yield for labeling biopolymers.

Formel: C₁₂H₁₄N₄O₅
Molecular Weight: 294,26 g/mol
CAS-Nummer: 88235-25-0
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR