NBD-X, succinimidyl ester ≥90%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: NBD-X, SE, Succinimidyl 6-(N-(7-nitrobenz-2-oxa-1,3-diazol-4-yl)amino)hexanoate

829
AATB829EA 460 EUR
AATB829
NBD-X, succinimidyl ester ≥90%
NBD-X, succinimidyl ester

Compared to NBD chloride and fluoride, NBD-X acid gives better yield for labeling biopolymers.

Formel: C₁₆H₁₇N₅O₇
Molecular Weight: 391,34 g/mol
CAS-Nummer: 145195-58-0
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR