Methylene blue NHS ester
Lieferant: AAT Bioquest
830
AATB830EA 410 EUR
AATB830
Methylene blue NHS ester
Methylene blue NHS ester
Methylene Blue (MB) is widely used as a redox indicator in nucleic acid research.
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only