Methylene blue maleimide
Lieferant: AAT Bioquest
831
AATB831EA 271 EUR
AATB831
Methylene blue maleimide
Methylene blue maleimide
Methylene Blue (MB) is widely used as a redox indicator in nucleic acid research.
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only