Methylene blue alkyne
Lieferant: AAT BIOQUEST
833
AATB833EA
271
EUR
AATB833
Methylene blue alkyne
Methylene blue alkyne
Methylene Blue (MB) is widely used as a redox indicator in nucleic acid research.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only