Pyranine 95

Lieferant: AAT BIOQUEST
Achtung

Synonyme: HPTS, 8-Hydroxy-1,3,6-pyrenetrisulphonic acid trisodium salt, Japan Green No. 204, Sodium 8-hydroxypyrene-1,3,6-trisulphonate, Trisodium 8-hydroxypyrene-1,3,6-trisulphonate

85
AATB85EA 120 EUR
AATB85
Pyranine 95
Pyranine

8-Hydroxypyrene-1,3,6-trisulfonic acid, trisodium salt [HPTS] is the cell-impermeable fluorescent pH indicator used for slightly acidic pH range (pKa = 6.4).

Formel: C₁₆H₇Na₃O₁₀S₃
Molecular Weight: 524.371 g/mol
MDL: MFCD00037575
CAS-Nummer: 6358-69-6
ADR: 5 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR