Rhodamine 110 ≥95%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: Rhodamine 110 chloride, 2-(6-Amino-3-imino-3H-xanthen-9-yl)benzoic acid hydrochloride

86
AATB86EA 230 EUR
AATB86
Rhodamine 110 ≥95%
Rhodamine 110

Rhodamine 110 (R110) is the precursor for a variety of R110-based enzyme substrates.

Formel: C₂₀H₁₅ClN₂O₃
Molecular Weight: 366,801 g/mol
MDL: MFCD00042009
CAS-Nummer: 13558-31-1
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR