Texas Red TCO fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

899
AATB899EA 292 EUR
AATB899
Texas Red TCO fluoreszierende Probe
Texas Red TCO

The tetrazine-trans-cyclooctene (TCO) ligation constitutes a non-toxic biomolecule labeling method of unparalleled speed.


  • Stable conjugation
  • Non-toxic labeling

Molecular Weight: 815,01 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=06ba1ad24ae8d2df13e9bff5f0c15cd9b5839e72&vid=99c6eac5bdab1c759be625e8ed2f264e6e61228f&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR