Cresyl violet perchlorate ≥90%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: Cresyl violet perchlorate, laser grade, 5-iminobenzo[a]phenoxazin-9-amine, perchloric acid

90
AATB90EA 120 EUR
AATB90
Cresyl violet perchlorate ≥90%
Cresyl violet perchlorate

It is used for staining Nissl substances in nerve cells and for bulk staining of nerve tissue.

Formel: C₁₆H₁₂ClN₃O₅
Molecular Weight: 361,738 g/mol
MDL: MFCD00012671
CAS-Nummer: 41830-80-2
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR