Cy3 trans-cyclooctene fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
Cy3 TCO
900
AATB900EA
307
EUR
AATB900
Cy3 trans-cyclooctene fluoreszierende Probe
Cy3 trans-cyclooctene
The tetrazine-trans-cyclooctene (TCO) ligation constitutes a non-toxic biomolecule labeling method of unparalleled speed.
- Stable conjugation
- Non-toxic labeling
Molecular Weight:
940,26 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...