Fluorescein aldehyde [5-FAM aldehyde]
Lieferant: AAT BIOQUEST
Fluorescein aldehyde is a reactive fluorescent dye that can react with an amine, hydrazine or hydroxylamine.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only