Cy5 trans-cyclooctene fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: Cy5 TCO

901
AATB901EA 307 EUR
AATB901
Cy5 trans-cyclooctene fluoreszierende Probe
Cy5 trans-cyclooctene

The tetrazine-trans-cyclooctene (TCO) ligation constitutes a non-toxic biomolecule labeling method of unparalleled speed.


  • Stable conjugation
  • Non-toxic labeling

Molecular Weight: 1161,58 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a7adf0fc68d82b6afcb464be53bc8ad4dd2ba58&vid=0cebfcec7570c9f6b572a9785634b7b567b076d7&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR