XFD488 aldehyde zur Proteinmarkierung

Lieferant: AAT BIOQUEST
Achtung

9015
AATB9015EA 586 EUR
AATB9015
XFD488 aldehyde zur Proteinmarkierung
XFD488 aldehyde

XFD488 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 488 (Alexa Fluor® is the trademark of ThermoFisher).


  • Reactive with amine, hydrazine, or hydroxylamine
  • Tolerates acidic pH
  • Detectable with the FITC channel

Molecular Weight: 750,75 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=72f9198607647fad7ea304228ffaaaa4f6a2ec67&vid=ea88543452fe7b66af749ebc0dfdae8ef8948670&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR