XFD488 aldehyde [Same Structure to Alexa Fluor™ 488 aldehyde]
Lieferant: AAT BIOQUEST
9016
AATB9016EA 429 EUR
AATB9016
XFD488 aldehyde [Same Structure to Alexa Fluor™ 488 aldehyde]
XFD488 aldehyde
XFD488 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 488 (Alexa Fluor® is the trademark of ThermoFisher).
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only