Cy5 tetrazine fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

902
AATB902EA 307 EUR
AATB902
Cy5 tetrazine fluoreszierende Probe
Cy5 tetrazine

The tetrazine-trans-cyclooctene (TCO) ligation constitutes a non-toxic biomolecule labeling method of unparalleled speed.


  • Stable conjugation
  • Non-toxic labeling

Molecular Weight: 1136,5 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=941a0a66b3537e9b9f26dfcee961e835740c820a&vid=d0dec1561f2ea0fbbaddb74cd690d3fd2b419580&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR