Cy5.5 tetrazine
Lieferant: AAT BIOQUEST
904
AATB904EA
309
EUR
AATB904
Cy5.5 tetrazine
Cy5.5 tetrazine
Cy5.5 tetrazine is an excellent building block that can be readily used to label TCO-modified biological molecules for NIR fluorescence imaging and other fluorescence-based biochemical analysis.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only