Cy3 tetrazine fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
910
AATB910EA
307
EUR
AATB910
Cy3 tetrazine fluoreszierende Probe
Cy3 tetrazine
The tetrazine-trans-cyclooctene (TCO) ligation constitutes a non-toxic biomolecule labeling method of unparalleled speed.
- Stable conjugation
- Non-toxic labeling
Molecular Weight:
908,07 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...