DBCO-Cy5 fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
923
AATB923EA
152
EUR
AATB923
DBCO-Cy5 fluoreszierende Probe
DBCO-Cy5
This azadibenzocyclooctyne-cyanine dye derivative is a versatile labeling reagent for detection of azide containing molecules or compounds.
- Copper-free reaction
- Azide reactive
Molecular Weight:
1211,6 g/mol |
CAS-Nummer:
1564286-24-3 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...