Cy3B acid fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
940
AATB940EA
769
EUR
AATB940
Cy3B acid fluoreszierende Probe
Cy3B acid
Cy3® dye is one of the most common cyanine dyes used for labeling proteins, nucleic acids and other biological molecules.
- High fluorescence quantum yield
- Improved photostability
Molecular Weight:
560,67 g/mol |
CAS-Nummer:
228272-69-3 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...