Cy3B maleimide
Lieferant: AAT BIOQUEST
942
AATB942EA 387 EUR
AATB942
Cy3B maleimide
Cy3B maleimide
Cy3® dye is one of the most common cyanine dyes used for labeling proteins, nucleic acids and other biological molecules.
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only