Indocyaningrün ≥95%

Lieferant: AAT Bioquest
Achtung

91 96
AATB91EA 460 EUR
AATB91 AATB96
Indocyaningrün ≥95%
Indocyanine green

Indocyanine green (ICG) is a cyanine dye used in medical diagnostics.

Formel: C₄₃H₄₇N₂NaO₆S₂
MDL: MFCD00013078
CAS-Nummer: 3599-32-4
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e512f181aaa9b4841ceb4424b30925b04e6e09f8&vid=ec59cd880ff67badb50a7e9b0b8a7fb649f03517&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR