6-TAMRA azide ≥95%
Lieferant: AAT Bioquest
6-TAMRA aside is an excellent alkyne-reactive fluorescent reagent for labeling alkyne-containing biological molecules through the well known click chemistry.
- Bright Fluorescence
- Click Chemistry Conjugation Reagent
Bitte beachten For research use only.