ICG azide fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
985
AATB985EA
307
EUR
AATB985
ICG azide fluoreszierende Probe
ICG azide
Indocyanine green (ICG) is a tricarbocyanine-type dye with NIR-absorbing properties (peak absorption around 780 nm) and emission maximum at ~800 nm.
- Low autofluorescence
- NIR fluorescence
Molecular Weight:
799,05 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...