ICG alkyne fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

986
AATB986EA 307 EUR
AATB986
ICG alkyne fluoreszierende Probe
ICG alkyne

Indocyanine green (ICG) is a tricarbocyanine-type dye with NIR-absorbing properties (peak absorption around 780 nm) and emission maximum at ~800 nm.


  • Low autofluorescence
  • NIR fluorescence

Molecular Weight: 768.03 g/mol
CAS-Nummer: 1622335-40-3
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=297c38972ec87c70176615b01b43a48163c2f80f&vid=17f69968c39f2fe32db2fe0f71fff9cc97b3fd86&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR