ICG hydrazide fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

987
AATB987EA 460 EUR
AATB987
ICG hydrazide fluoreszierende Probe
ICG hydrazide

Indocyanine green (ICG) is a tricarbocyanine-type dye with NIR-absorbing properties (peak absorption around 780 nm) and emission maximum at ~800 nm.


  • Low autofluorescence
  • NIR fluorescence

Molecular Weight: 973,04 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=504909f815bf16bb0bdf9b9462a82db2526a6e14&vid=5fe77bc34d54af1d52d94f43883822f47f3b59bd&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR