ICG-TFP ester fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

990
AATB990EA 460 EUR
AATB990
ICG-TFP ester fluoreszierende Probe
ICG-TFP ester

Indocyanine green (ICG) is a cyanine dye used in medical diagnostics.


  • Low autofluorescence
  • NIR fluorescence

Molecular Weight: 981,06 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2997c7fcb0a65c81edffb5cf2886d92aeb85e762&vid=c01874de8a28f8400a6afcec091bed492c79d3a3&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR