iFluor® 488, Alkin fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

999
AATB999EA 307 EUR
AATB999
iFluor® 488, Alkin fluoreszierende Probe
iFluor®

Although FITC is still the most popular fluorescent labeling dye for preparing green fluorescent bioconjugates, there are certain limitations with FITC, such as severe photobleaching for microscope imaging and pH-sensitive fluorescence.


  • pH-insensitive
  • Azide reactive
  • High photostability
  • Brighter signal

ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2e12400d8cc05c1a62530e3036cfa0069a123ce&vid=6baadb1a10fd4664a71904dd4913623105e13ce3&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR