25-Hydroxyvitamin D3 ≥98%

Lieferant: Abcam
Gefahr

Synonyme: Calcifediol, (R,Z)-3-(2-((1R,3aS,7aR,E)-1-((R)-6-hydroxy-6-methylheptan-2-yl)-7a-methyloctahydro-4H-inden-4-ylidene)ethylidene)-4-methylenecyclohexan-1-ol

AB142766-5
ABCAAB142766-5EA 953 EUR
ABCAAB142766-5
25-Hydroxyvitamin D3 ≥98%
25-Hydroxyvitamin D3

The major circulating metabolite of vitamin D3 (Cholecalciferol). Calcifediol also has mineralisng properties.

Formel: C₂₇H₄₄O₂
Molecular Weight: 400,6 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00867077
CAS-Nummer: 19356-17-3
UN: 2811
ADR: 6.1,II

Bestellen

Spezifikation Testergebnisse

Purity >98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR