Carbonic Anhydrase (CA) Inhibitor Screening Kit (Colorimetric)
Lieferant: Abcam
AB283387-100
ABCAAB283387-100EA 875 EUR
ABCAAB283387-100
Carbonic Anhydrase (CA) Inhibitor Screening Kit (Colorimetric)
Tests Enzymtests
It is simple and can be used to identify and characterize CA inhibitors in a high-throughput format.
  • Simple, highly sensitive, high-throughput compatible
  • Rapid screening of Carbonic Anhydrase ligands
  • Kit includes Acetazolamide, a CA inhibitor, and a stable, recombinant human CA
Carbonic anhydrases (CA; EC: 4.2.1.1) are zinc enzymes present in both prokaryotes and eukaryotes. They efficiently catalyse the reversible hydration of CO2 to bicarbonate. Their important patho-physiological roles in respiration, pH and CO2 homeostasis, secretion, gluconeogenesis, ureagenesis etc. make it an important drug target. CA inhibition in different organs can result in different effects. Inhibition of CA helps in different disease treatment procedures; for example, it can help in the treatment of Glaucoma, urine alkalisation, managing epilepsy, sleep apnea, acute mountain sickness etc. BioVision’s CA Inhibitor Screening Kit can be used to screen for potent inhibitors of CA activity. It utilises the esterase activity of an active CA on an ester substrate which releases a chromogenic product. The released product can be easily quantified using an absorbance microplate reader. In the presence of a CA specific inhibitor, the enzyme loses its activity which results in decrease of absorbance. This assay kit is simple and can be used to identify and characterize CA inhibitors in a high-throughput format. Here, we have used the specific inhibitor Acetazolamide, a potent inhibitor of CA I, CA II, CA IV, CA IX, CA XII etc. except CA III