Doxycycline ELISA Kit, Inc.
Lieferant: Abcam
A competitive ELISA kit for the quantitative measurement of doxycycline in tissue, honey, milk, serum.
- Detection Range: 0,2 to 5,4 ppb
- Sensitivity: <0,2 ppb
- Detection limitation: 2 ppb for serum, 5 ppb tissue, honey and milk, 10 ppb for milk
- Cross reaction: Doxycycline 100%, tetracycline, minocycline 100%, chlortetracycline 180%, oxytetracyline 40%
- Easy, convenient and time-saving
Doxycycline is A synthetic derivative of oxytetracycline. Acts as a broad spectrum antibiotic and matrix metallo-proteinases (MMP) inhibitor.
Useful in studies involving wound healing and tissue remodeling. Common side effects include diarrhea, nausea, vomiting, a red rash, and an increased risk of a sunburn. Use during pregnancy or in young children may result in permanent problems with the teeth including changes in their colour.
Standard techniques/instruments (HPLC or GC-MS) are utilized to detect doxycycline. However, these techniques are complex, expensive, laborious, and time-consuming. Immunoassay techniques, such as ELISAs are commonly preferred as a simple, reliable and rapid method for the quantification of doxycycline in various samples.
Doxycycline ELISA Kit is a competitive-based ELISA that can be used for the determination of this antibiotic in tissue, honey, milk and serum.
Bitte beachten
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...