Hepcidin (Rat) ELISA Kit

Lieferant: Abcam

AB285233-96
ABCAAB285233-96EA 1290 EUR
ABCAAB285233-96
Hepcidin (Rat) ELISA Kit
Tests ELISAs

A sandwich ELISA kit for the qualitative detect Hepcidin in rat serum or plasma.


  • Sensitivity: <9,375 pg/ml, detection Range: 15,625 to 1000 pg/ml, no significant cross-reactivity or interference was observed
  • Easy, convenient and time-saving method to measure the level of Hepcidin in serum, plasma, tissue homogenates, culture supernatants and other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter-Assay: CV <10%


Hepcidin is a liver-produced hormone that constitutes the main circulating regulator of iron absorption and distribution across tissues. Acts by promoting endocytosis and degradation of ferroportin, leading to the retention of iron in iron-exporting cells and decreased flow of iron into plasma. Controls the major flows of iron into plasma: absorption of dietary iron in the intestine, recycling of iron by macrophages, which phagocytose old erythrocytes and other cells, and mobilization of stored iron from hepatocytes. BioVision's Hepcidin ELISA kit is a sandwich ELISA assay for the quantitative measurement of Hepc in rat serum, plasma and cell culture supernatants. The density of color is proportional to the amount of Hepcidin captured from the samples.


Bitte beachten

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.


Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR