Estrogen ELISA Kit
Lieferant: Abcam
A competitive ELISA kit for the quantitative measurement of estrogen in biological fluids.
- Detection range: 15,625 to 1000 pg/ml
- Sensitivity: <9,375 pg/ml
- Assay precision: Intra-assay: CV <8%; Inter-Assay: CV <10%
- No significant cross-reactivity or interference was observed
Easy, convenient and time-saving method to measure the level of estrogen in serum, plasma, tissue homogenates, culture supernatants and other biological fluids.
Estrogen is the primary female sex hormone. It is responsible for the development and regulation of the female reproductive system and secondary sex characteristics.
Estrogen may also refer to any substance, natural or synthetic, that mimics the effects of the natural hormone. The steroid 17β-estradiol is the most potent and prevalent endogenous estrogen, although several metabolites of estradiol also have estrogenic hormonal activity.
Synthetic estrogens are used as part of some oral contraceptives, in estrogen replacement therapy for postmenopausal women, and in hormone replacement therapy for trans women. Estrogen ELISA kit is a competitive ELISA assay for the quantitative measurement of estrogen in human serum, plasma and cell culture supernatants.
The density of colour is proportional to the amount of estrogen captured from the samples.
Bitte beachten
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...