QuickDetect™ 1-3-β-D-glucan (Human) ELISA Kit
Lieferant: Abcam
AB287791-96
ABCAAB287791-96EA 954 EUR
ABCAAB287791-96
QuickDetect™ 1-3-β-D-glucan (Human) ELISA Kit
Tests ELISAs
A rapid ELISA kit for quantify of 1-3-β-D-glucan in human serum, plasma, urine, cell culture samples.
  • Easy, convenient and time-saving method to measure the level of 1-3-β-D-glucan in human Serum, plasma, urine, cell culture samples, biological fluid
  • Detection Range: 0,8 to 50 pg/ml
  • Sensitivity: <0,1 pg/ml
β-Glucans (beta-glucans) comprise a group of β-D-glucose polysaccharides naturally occurring in the cell walls of cereals, bacteria, and fungi, with significantly differing physicochemical properties dependent on source. Typically, β-glucans form a linear backbone with 1-3 β-glycosidic bonds but vary with respect to molecular mass, solubility, viscosity, branching structure, and gelation properties, causing diverse physiological effects in animals. BioVision’s 1-3-β-DG ELISA kit is a sandwich ELISA assay for the quantitative measurement of Human 1-3-β-DG in serum, plasma and cell culture supernatants in 90 minutes. The density of colour is proportional to the amount of Human 1-3-β-DG captured from the samples.
Bitte beachten
All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.