Arachidonic Acid (AA) ELISA Kit

Lieferant: Abcam

AB287798-96
ABCAAB287798-96EA 1240 EUR
ABCAAB287798-96
Arachidonic Acid (AA) ELISA Kit
Tests ELISAs

A competitive ELISA kit for the quantitative measurement of arachidonic acid in biological fluids.


  • Detection range: 78,125 to 5000 ng/ml
  • Sensitivity: <69,74 ng/ml
  • Assay precision: Intra-assay: CV <8%; inter-assay: CV <10%
  • No significant cross-reactivity or interference was observed


Easy, convenient and time-saving method to measure the level of arachidonic acid in serum, plasma, tissue homogenates, culture supernatants and other biological fluids.


Arachidonic acid (AA, ARA) is a polyunsaturated omega-6 fatty acid 20:4(ω-6). It is structurally related to the saturated arachidic acid found in cupuaçu butter. It is a precursor to a large family of eicosanoids. Activator for several protein kinase C isotypes.


Arachidonic acid ELISA kit is a competitive ELISA assay for the quantitative measurement of arachidonic acid in serum, plasma and cell culture supernatants. The density of colour is proportional to the amount of arachidonic acid captured from the samples.


Bitte beachten

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.


Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR