Human ADAM10 ELISA Kit
Lieferant: Abcam
Human ADAM10 ELISA kit is a single-wash 90-min Simplestep used to quantify Human ADAM10 with a sensitivity of 5.38 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader .