4-(Brommethyl)benzolsulfonylchlorid 95%
Lieferant: ABCR
|
Gefahr
|
Synonyme:
4-(Brommethyl)benzolsulfonsäurechlorid, 4-(Brommethyl)benzol-1-sulfonylchlorid, 4-(Brommethyl)benzolsulfochlorid
10120.0025
ABCR10120.0025EA
0
EUR
ABCR10120.0025
4-(Brommethyl)benzolsulfonylchlorid 95%
4-(Brommethyl)benzolsulfonylchlorid
Formel:
C₇H₆BrClO₂S Molecular Weight: 269,55 g/mol Schmelzpunkt: 72…75 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00156129 CAS-Nummer: 66176-39-4 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...